missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-chloropyrimidine-5-carboxylate, 95%, Thermo Scientific™

Catalog Number 15219906
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
89793-12-4
C7H7ClN2O2
186.6
IEMKQRSOAOPKRJ-UHFFFAOYSA-N
2-chloro-pyrimidine-5-carboxylic acid ethyl ester, 2-chloro-5-ethoxycarbonyl pyrimidine, ethyl-2-chloropyrimidine-5-carboxylate, ethyl 2-chloro-pyrimidine-5-carboxylate, 5-pyrimidinecarboxylic acid, 2-chloro-, ethyl ester, ethyl2-chloropyrimidine-5-carboxylate, acmc-209r2w, ksc663i8r, ethyl 2-chloranylpyrimidine-5-carboxylate, 2-chloro-5-pyrimidinecarboxylic acid ethyl ester
10487815
ethyl 2-chloropyrimidine-5-carboxylate
CCOC(=O)C1=CN=C(N=C1)Cl
This item is not returnable. View return policy
89793-12-4
C7H7ClN2O2
186.6
IEMKQRSOAOPKRJ-UHFFFAOYSA-N
2-chloro-pyrimidine-5-carboxylic acid ethyl ester, 2-chloro-5-ethoxycarbonyl pyrimidine, ethyl-2-chloropyrimidine-5-carboxylate, ethyl 2-chloro-pyrimidine-5-carboxylate, 5-pyrimidinecarboxylic acid, 2-chloro-, ethyl ester, ethyl2-chloropyrimidine-5-carboxylate, acmc-209r2w, ksc663i8r, ethyl 2-chloranylpyrimidine-5-carboxylate, 2-chloro-5-pyrimidinecarboxylic acid ethyl ester
10487815
ethyl 2-chloropyrimidine-5-carboxylate
CCOC(=O)C1=CN=C(N=C1)Cl
Melting Point 57°C to 58°C
Assay Percent Range 94% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 186.6
Percent Purity 95%
Chemical Name or Material Ethyl 2-chloropyrimidine-5-carboxylate
compliance-icons
Product Identifier
  • Ethyl 2-chloropyrimidine-5-carboxylate, 95%
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'