Läs mer
Ethyl 2-Cyanopropanoate, TRC
Produktkod. 30376627
Change view
Kvantitet:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30376627 5 g
30369541 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30376627 Leverantör Toronto Research Chemicals Leverantörsnummer TRCE9077605G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Ethyl 2-Cyanopropanoate
CAS 1572-99-2
Molekylformel C21 H26 N2 O
Kvantitet 5 g
Synonym 2-Cyanopropanoic Acid Ethyl Ester, Ethyl 2-Cyanopropionate, Ethyl alpha-Cyanopropionate, NSC 68508
InChI formel InChI=1S/C21H26N2O/c1-16(13-22(2)3)14-23-20-7-5-4-6-18(20)9-10-19-12-17(15-24)8-11-21(19)23/h4-8,11-12,15-16H,9-10,13-14H2,1-3H3
LEDER CC(CN(C)C)CN1c2ccccc2CCc3cc(C=O)ccc13
IUPAC-namn 11-[3-(dimethylamino)-2-methylpropyl]-5,6-dihydrobenzo[b][1]benzazepine-3-carbaldehyde
Molekylvikt (g/mol) 322.444
Formel vikt 322.205