Läs mer
Ethyl 2-Cyclopropyl-2-hydroxyacetate, TRC
Produktkod. 30012183
Change view
Kvantitet:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30012183 100 mg
30005797 50 mg
30006301 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30012183 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB125348100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1185387-66-9
Molekylformel C7 H12 O3
Molekylvikt (g/mol) 144.168
IUPAC-namn ethyl 2-cyclopropyl-2-hydroxyacetate
LEDER CCOC(=O)C(O)C1CC1
Specifikationer
Kemiskt namn eller material Ethyl 2-Cyclopropyl-2-hydroxyacetate
Kvantitet 100 mg
InChI formel InChI=1S/C7H12O3/c1-2-10-7(9)6(8)5-3-4-5/h5-6,8H,2-4H2,1H3
Formel vikt 144.079