Läs mer
Ethyl 2-Ethoxy-1-((2'-(5-oxo-2,5-dihydro-1,2,4-oxadiazol-3-yl)-[1,1'-biphenyl]-4-yl)methyl)-1H-benzo[d]imidazole-7-carboxylate, TRC
Produktkod. 30137588
Change view
Kvantitet:
100 mg
250 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30137588 250 mg
30138755 500 mg
30007597 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30137588 Leverantör Toronto Research Chemicals Leverantörsnummer TRCE678645250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1403474-70-3
Molekylformel C27 H24 N4 O5
Molekylvikt (g/mol) 484.502
Synonym Ethyl 1-[[2'-(2,5-Dihydro-5-oxo-1,2,4-oxadiazol-3-yl)[1,1'-biphenyl]-4-yl]methyl]-2-ethoxy-1H-benzimidazole-7-carboxylate, Azilsartan Kamedoxomil Impurity, Azilsartan Impurity;
IUPAC-namn ethyl 2-ethoxy-3-[[4-[2-(5-oxo-2H-1,2,4-oxadiazol-3-yl)phenyl]phenyl]methyl]benzimidazole-4-carboxylate
LEDER CCOC(=O)c1cccc2nc(OCC)n(Cc3ccc(cc3)c4ccccc4C5=NC(=O)ON5)c12
Specifikationer
Kemiskt namn eller material Ethyl 2-Ethoxy-1-((2'-(5-oxo-2,5-dihydro-1,2,4-oxadiazol-3-yl)-[1,1'-biphenyl]-4-yl)methyl)-1H-benzo[d]imidazole-7-carboxylate
Kvantitet 250 mg
InChI formel InChI=1S/C27H24N4O5/c1-3-34-25(32)21-10-7-11-22-23(21)31(26(28-22)35-4-2)16-17-12-14-18(15-13-17)19-8-5-6-9-20(19)24-29-27(33)36-30-24/h5-15H,3-4,16H2,1-2H3,(H,29,30,33)
Formel vikt 484.1747