Läs mer
Ethyl-2-ethoxy-1-[[(2'-cyanobiphenyl-4-yl) methyl] benzimidazole]-7-carboxylate, TRC
Produktkod. 30376933
Change view
Kvantitet:
500 mg
Produktkod. Kvantitet
30376933 500 mg
1 options
Produktkod. 30376933 Leverantör Toronto Research Chemicals Leverantörsnummer TRCE900068500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Ethyl 1-[(2'-Cyanobiphenyl-4-yl)methyl]-2-ethoxy-1H-benzimidazole-7-carboxylate
CAS 139481-41-7
Molekylformel C26 H23 N3 O3
Kvantitet 500 mg
Synonym 1H-Benzimidazole-7-carboxylic acid, 1-[(2'-cyano[1,1'-biphenyl]-4-yl)methyl]-2-ethoxy-, ethyl ester, 1-[(2'-Cyano-1,1'-biphenyl-4-yl)methyl]-2-ethoxy-7-benzimidazolecarboxylic acid ethyl ester, Ethyl 1-[(2'-cyanobiphenyl-4-yl)-methyl]-2-ethoxybenzimidazole-7-carboxylate, Ethyl 1-[(2'-Cyanobiphenyl-4-yl)methyl]-2-ethoxy-1H-benzimidazole-7-carboxylate
InChI formel InChI=1S/C26H23N3O3/c1-3-31-25(30)22-10-7-11-23-24(22)29(26(28-23)32-4-2)17-18-12-14-19(15-13-18)21-9-6-5-8-20(21)16-27/h5-15H,3-4,17H2,1-2H3
LEDER CCOC(=O)c1cccc2nc(OCC)n(Cc3ccc(cc3)c4ccccc4C#N)c12
IUPAC-namn ethyl 3-[[4-(2-cyanophenyl)phenyl]methyl]-2-ethoxybenzimidazole-4-carboxylate
Molekylvikt (g/mol) 425.48
Formel vikt 425.1739