Läs mer
Ethyl 2-Ethylhexanoate, TRC
Produktkod. 30375068
Change view
Kvantitet:
1 g
Produktkod. Kvantitet
30375068 1 g
1 options
Produktkod. 30375068 Leverantör Toronto Research Chemicals Leverantörsnummer TRCE9187401G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Ethyl 2-Ethylhexanoate
CAS 2983-37-1
Molekylformel C10H20O2
Kvantitet 1 g
Synonym Ethyl alpha-Ethylcaproate, Irotyl
InChI formel InChI=1S/C10H20O2/c1-4-7-8-9(5-2)10(11)12-6-3/h9H,4-8H2,1-3H3
LEDER CCCCC(CC)C(=O)OCC
IUPAC-namn ethyl 2-ethylhexanoate
Molekylvikt (g/mol) 172.26
Formel vikt 172.1463