Läs mer
Ethyl 2-Fluoropropionate, TRC
Produktkod. 30371510
Change view
Kvantitet:
5 g
Produktkod. Kvantitet
30371510 5 g
1 options
Produktkod. 30371510 Leverantör Toronto Research Chemicals Leverantörsnummer TRCE9179655G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Ethyl 2-Fluoropropionate
CAS 349-43-9
Molekylformel C5H9FO2
Kvantitet 5 g
Synonym 2-Fluoropropanoic Acid Ethyl Ester, 2-Fluoropropionic Acid Ethyl Ester, Ethyl 2-Fluoropropanoate, Ethyl alpha-Fluoropropionate, NSC 102760
InChI formel InChI=1S/C5H9FO2/c1-3-8-5(7)4(2)6/h4H,3H2,1-2H3
LEDER CCOC(=O)C(C)F
IUPAC-namn ethyl 2-fluoropropanoate
Molekylvikt (g/mol) 120.12
Formel vikt 120.0587