Läs mer
Ethyl 2-Hydroxyphenylacetate, TRC
Produktkod. 30379330
Change view
Kvantitet:
1 g
Produktkod. Kvantitet
30379330 1 g
1 options
Produktkod. 30379330 Leverantör Toronto Research Chemicals Leverantörsnummer TRCE9190701G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Ethyl 2-Hydroxyphenylacetate
CAS 41873-65-8
Molekylformel C10 H12 O3
Kvantitet 1 g
Synonym Benzeneacetic acid, 2-hydroxy-, ethyl ester, 2-(2-Hydroxyphenyl)acetic acid ethyl ester, Ethyl (2-hydroxyphenyl)acetate, Ethyl 2-hydroxybenzeneacetate
InChI formel InChI=1S/C10H12O3/c1-2-13-10(12)7-8-5-3-4-6-9(8)11/h3-6,11H,2,7H2,1H3
LEDER CCOC(=O)Cc1ccccc1O
IUPAC-namn ethyl 2-(2-hydroxyphenyl)acetate
Molekylvikt (g/mol) 180.2
Formel vikt 180.0786