Learn More
Ethyl 2-mercaptoacetate, 99%, Thermo Scientific Chemicals
Catalog Number 10040930
1577.91 SEK valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
623-51-8
C4H8O2S
120.17
MFCD00004874
PVBRSNZAOAJRKO-UHFFFAOYSA-N
ethyl thioglycolate, ethyl mercaptoacetate, ethyl 2-mercaptoacetate, mercaptoacetic acid ethyl ester, ethyl thioglycollate, acetic acid, mercapto-, ethyl ester, thioglycolic acid ethyl ester, ethyl mercaptoacetic acid, usaf ek-2070, acetic acid, 2-mercapto-, ethyl ester
12185
ethyl 2-sulfanylacetate
CCOC(=O)CS
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
623-51-8
C4H8O2S
120.17
MFCD00004874
PVBRSNZAOAJRKO-UHFFFAOYSA-N
ethyl thioglycolate, ethyl mercaptoacetate, ethyl 2-mercaptoacetate, mercaptoacetic acid ethyl ester, ethyl thioglycollate, acetic acid, mercapto-, ethyl ester, thioglycolic acid ethyl ester, ethyl mercaptoacetic acid, usaf ek-2070, acetic acid, 2-mercapto-, ethyl ester
12185
ethyl 2-sulfanylacetate
CCOC(=O)CS
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Melting Point -80°C
Color Colorless
Density 1.0900g/mL
Boiling Point 54°C (12.0 mmHg)
Flash Point 47°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula HSCH2CO2C2H5
Refractive Index 1.4560 to 1.459
Quantity 500 g
Beilstein 03,255
Specific Gravity 1.09
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in ethanol,acetone
Viscosity 1.2 mPa.s (20°C)
Formula Weight 120.17
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Ethyl 2-mercaptoacetate, 99%