Läs mer
Ethyl [2-Methyl-3-(chlorosulfonyl)phenoxy]acetate, TRC
Produktkod. 30373681
Change view
Kvantitet:
5 g
Produktkod. Kvantitet
30373681 5 g
1 options
Produktkod. 30373681 Leverantör Toronto Research Chemicals Leverantörsnummer TRCE9250705G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Ethyl [2-Methyl-3-(chlorosulfonyl)phenoxy]acetate
CAS 91427-62-2
Molekylformel C11 H13 Cl O5 S
Kvantitet 5 g
Synonym Acetic acid, 2-[4-(chlorosulfonyl)-2-methylphenoxy]-, ethyl ester, Acetic acid, [4-(chlorosulfonyl)-2-methylphenoxy]-, ethyl ester (9CI), Acetic acid, [[4-(chlorosulfonyl)-o-tolyl]oxy]-, ethyl ester (7CI), (4-Chlorosulfonyl-2-methylphenoxy)acetic acid ethyl ester, Ethyl 2-[4-(chlorosulfonyl)-2-methylphenoxy]acetate, Ethyl [2-Methyl-3-(chlorosulfonyl)phenoxy]acetate
InChI formel InChI=1S/C11H13ClO5S/c1-3-16-11(13)7-17-9-5-4-6-10(8(9)2)18(12,14)15/h4-6H,3,7H2,1-2H3
LEDER CCOC(=O)COc1cccc(c1C)S(=O)(=O)Cl
IUPAC-namn ethyl 2-(3-chlorosulfonyl-2-methylphenoxy)acetate
Molekylvikt (g/mol) 292.74
Formel vikt 292.0172