missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-methyl-4-phenyl-1,3-thiazole-5-carboxylate, 97%, Thermo Scientific™

Catalog Number p-3486096
Click to view available options
Quantity:
250 mg
1 g
10 g
This item is not returnable. View return policy
32043-95-1
C13H13NO2S
247.31
MFCD07346320
UHLMXNFHHFDVPW-UHFFFAOYSA-N
ethyl 2-methyl-4-phenylthiazole-5-carboxylate, 2-methyl-4-phenylthiazole-5-carboxylic acid ethyl ester, 2-methyl-4-phenyl-thiazole-5-carboxylic acid ethyl ester, 5-thiazolecarboxylicacid, 2-methyl-4-phenyl-, ethyl ester
7131013
ethyl 2-methyl-4-phenyl-1,3-thiazole-5-carboxylate
CCOC(=O)C1=C(N=C(C)S1)C1=CC=CC=C1
This item is not returnable. View return policy
32043-95-1
C13H13NO2S
247.31
MFCD07346320
UHLMXNFHHFDVPW-UHFFFAOYSA-N
ethyl 2-methyl-4-phenylthiazole-5-carboxylate, 2-methyl-4-phenylthiazole-5-carboxylic acid ethyl ester, 2-methyl-4-phenyl-thiazole-5-carboxylic acid ethyl ester, 5-thiazolecarboxylicacid, 2-methyl-4-phenyl-, ethyl ester
7131013
ethyl 2-methyl-4-phenyl-1,3-thiazole-5-carboxylate
CCOC(=O)C1=C(N=C(C)S1)C1=CC=CC=C1
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 250 mg
Formula Weight 247.32
Percent Purity 97%
Chemical Name or Material Ethyl 2-methyl-4-phenyl-1, 3-thiazole-5-carboxylate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'