missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-morpholino-1,3-thiazole-4-carboxylate, 97%, Thermo Scientific™

Catalog Number p-3541048
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
126533-95-7
C10H14N2O3S
242.293
MFCD09702412
OINWWRRVBHJAKO-UHFFFAOYSA-N
ethyl 2-morpholin-4-yl-1,3-thiazole-4-carboxylate, ethyl 2-4-morpholinyl thiazole-4-carboxylate, ethyl 2-morpholino-1,3-thiazole-4-carboxylate, ethyl 2-morpholinothiazole-4-carboxylate, 4-thiazolecarboxylicacid, 2-4-morpholinyl-, ethyl ester, acmc-1c0vg, 4-4-ethoxycarbonyl-1,3-thiazol-2-yl morpholine, 4-thiazolecarboxylicacid,2-4-morpholinyl-,ethyl ester, 2-morpholin-4-yl-thiazole-4-carboxylic acid ethyl ester
15053544
ethyl 2-morpholin-4-yl-1,3-thiazole-4-carboxylate
CCOC(=O)C1=CSC(=N1)N2CCOCC2
This item is not returnable. View return policy
126533-95-7
C10H14N2O3S
242.293
MFCD09702412
OINWWRRVBHJAKO-UHFFFAOYSA-N
ethyl 2-morpholin-4-yl-1,3-thiazole-4-carboxylate, ethyl 2-4-morpholinyl thiazole-4-carboxylate, ethyl 2-morpholino-1,3-thiazole-4-carboxylate, ethyl 2-morpholinothiazole-4-carboxylate, 4-thiazolecarboxylicacid, 2-4-morpholinyl-, ethyl ester, acmc-1c0vg, 4-4-ethoxycarbonyl-1,3-thiazol-2-yl morpholine, 4-thiazolecarboxylicacid,2-4-morpholinyl-,ethyl ester, 2-morpholin-4-yl-thiazole-4-carboxylic acid ethyl ester
15053544
ethyl 2-morpholin-4-yl-1,3-thiazole-4-carboxylate
CCOC(=O)C1=CSC(=N1)N2CCOCC2
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 242.3
Percent Purity 97%
Chemical Name or Material Ethyl 2-morpholino-1, 3-thiazole-4-carboxylate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'