Läs mer
Ethyl 2-Oxo-2-(2-oxotetrahydrofuran-3-yl)acetate, TRC
Produktkod. 30371283
Change view
Kvantitet:
250 mg
Produktkod. Kvantitet
30371283 250 mg
1 options
Produktkod. 30371283 Leverantör Toronto Research Chemicals Leverantörsnummer TRCE925880250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Ethyl 2-Oxo-2-(2-oxotetrahydrofuran-3-yl)acetate
CAS 42564-36-3
Molekylformel C8H10O5
Kvantitet 250 mg
Synonym Tetrahydro-alpha,2-dioxo-3-furanacetic Acid Ethyl Ester, (2-Hydroxyethyl)oxalacetic Acid gamma-Lactone Ethyl Ester
InChI formel InChI=1S/C8H10O5/c1-2-12-8(11)6(9)5-3-4-13-7(5)10/h5H,2-4H2,1H3
LEDER CCOC(=O)C(=O)C1CCOC1=O
IUPAC-namn ethyl 2-oxo-2-(2-oxooxolan-3-yl)acetate
Molekylvikt (g/mol) 186.16
Formel vikt 186.0528