Läs mer
Ethyl 2-Oxopentanoate, TRC
Produktkod. 30373787
Change view
Kvantitet:
2.5 g
Produktkod. Kvantitet
30373787 2.5 g
1 options
Produktkod. 30373787 Leverantör Toronto Research Chemicals Leverantörsnummer TRCE9258052.5G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Ethyl 2-Oxopentanoate
CAS 50461-74-0
Molekylformel C7H12O3
Kvantitet 2.5 g
Synonym 2-Oxopentanoic Acid Ethyl Ester, 2-Oxovaleric Acid Ethyl Ester, 2-Oxopentanoic Acid Ethyl Ester
InChI formel InChI=1S/C7H12O3/c1-3-5-6(8)7(9)10-4-2/h3-5H2,1-2H3
LEDER CCCC(=O)C(=O)OCC
IUPAC-namn ethyl 2-oxopentanoate
Molekylvikt (g/mol) 144.17
Formel vikt 144.0786