Läs mer
Ethyl (2R,4S)-4-Methylpipecolate, >90%, TRC
Produktkod. 30370975
Change view
Kvantitet:
10 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30370975 10 mg
30377525 25 mg
30372720 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30370975 Leverantör Toronto Research Chemicals Leverantörsnummer TRCE92463510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 79199-61-4
Molekylformel C9H17NO2
Molekylvikt (g/mol) 171.24
Synonym (2R,4S)-4-Methyl-2-piperidinecarboxylic Acid Ethyl Ester, (2R-cis)-4-Methyl-2-piperidinecarboxylic Acid Ethyl Ester
IUPAC-namn ethyl (2R,4S)-4-methylpiperidine-2-carboxylate
LEDER CCOC(=O)[C@H]1C[C@H](CCN1)C
Specifikationer
Kemiskt namn eller material Ethyl (2R,4S)-4-Methylpipecolate
Kvantitet 10 mg
InChI formel InChI=1S/C9H17NO2/c1-3-12-9(11)8-6-7(2)4-5-10-8/h7-8,10H,3-6H2,1-2H3/t7-,8+/m0/s1
Formel vikt 171.1259