missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Ethyl (2S)-2-[(3S,8aS)-3-Methyl-1,4-dioxooctahydropyrrolo[1,2-a]pyrazin-2-yl]-4-phenylbutanoate (Enalapril Diketopiperazine), Mikromol™
Beskrivning
Ethyl (2S)-2-[(3S,8aS)-3-Methyl-1,4-dioxooctahydropyrrolo[1,2-a]pyrazin-2-yl]-4-phenylbutanoate (Enalapril Diketopiperazine)
Specifikationer
Specifikationer
| Kemiskt namn eller material | Enalapril Diketopiperazine |
| CAS | 115729-52-7 |
| Analyt- eller komponentnamn | Enalapril Diketopiperazine |
| Frakt skick | Room Temperature |
| Molekylformel | C20 H26 N2 O4 |
| Certifikat och efterlevnad | ISO 17025 |
| Synonym | Enalapril Maleate Imp. D (EP), Enalapril Imp. D (EP), Ethyl (2S)-2-[(3S,8aS)-3-Methyl-1,4-dioxooctahydropyrrolo[1,2-a]pyrazin-2-yl]-4-phenylbutanoate, Enalapril Diketopiperazine, Enalapril Related Compound D, Enalapril Maleate Impurity D, Enalapril Impurity D |
| InChI formel | InChI=1S/C20H26N2O4/c1-3-26-20(25)17(12-11-15-8-5-4-6-9-15)22-14(2)18(23)21-13-7-10-16(21)19(22)24/h4-6,8-9,14,16-17H,3,7,10-13H2,1-2H3/t14-,16-,17-/m0/s1 |
| LEDER | CCOC(=O)[C@H](CCc1ccccc1)N2[C@@H](C)C(=O)N3CCC[C@H]3C2=O |
| IUPAC-namn | ethyl (2S)-2-[(3S,8aS)-3-methyl-1,4-dioxo-6,7,8,8a-tetrahydro-3H-pyrrolo[1,2-a]pyrazin-2-yl]-4-phenylbutanoate |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?