Läs mer
Ethyl 3-[[3-Amino-4-(methylamino)benzoyl]-(2-pyridyl)amino]propanoate (N-[3-Amino-4-(methylamino)benzoyl]-N-2-pyridinyl-beta-alanine Ethyl Ester), Mikromol
Produktkod. 30459339
Change view
Kvantitet:
25 mg
4 x 25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30459339 4 x 25 mg
30050093 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30459339 Leverantör Mikromol Leverantörsnummer MM3522.0100254AMP
Discover Mikromol- your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Ethyl 3-[[3-Amino-4-(methylamino)benzoyl]-(2-pyridyl)amino]propanoate (N-[3-Amino-4-(methylamino)benzoyl]-N-2-pyridinyl-beta-alanine Ethyl Ester), Mikromol
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material N-(3-Amino-4-(methylamino)benzoyl)-N-2-pyridinyl-Beta-alanine Ethyl Ester
CAS 212322-56-0
Analyt- eller komponentnamn N-(3-Amino-4-(methylamino)benzoyl)-N-2-pyridinyl-Beta-alanine Ethyl Ester
Frakt skick Room Temperature
Molekylformel C18 H22 N4 O3
Certifikat och efterlevnad ISO 17025
Synonym Ethyl 3-((3-amino-4-methylaminobenzoyl)-(pyridin-2-yl)-amino)propanoate, N-(3-amino-4-(methylamino)benzoyl)-N-2-pyridinyl-beta-Alanine ethyl ester
InChI formel InChI=1S/C18H22N4O3/c1-3-25-17(23)9-11-22(16-6-4-5-10-21-16)18(24)13-7-8-15(20-2)14(19)12-13/h4-8,10,12,20H,3,9,11,19H2,1-2H3
Lösningstyp Neat
LEDER CCOC(=O)CCN(C(=O)c1ccc(NC)c(N)c1)c2ccccn2
IUPAC-namn ethyl 3-((3-amino-4-(methylamino)benzoyl)-pyridin-2-ylamino)propanoate
Molekylvikt (g/mol) 342.39
Kvantitet 4 x 25 mg
Formel vikt 342.1692 g/mol