Läs mer
Ethyl 3-(3-(tert-Butyl)-4-hydroxy-5-methylphenyl)propanoate, TRC
Produktkod. 30180235
Change view
Kvantitet:
50 mg
250 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30180235 500 mg
30180086 50 mg
30180146 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30180235 Leverantör Toronto Research Chemicals Leverantörsnummer TRCE178240500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 40200-35-9
Molekylformel C16 H24 O3
Molekylvikt (g/mol) 264.36
Synonym Benzenepropanoic acid, 3-(1,1-dimethylethyl)-4-hydroxy-5-methyl-, ethyl ester, Ethyl 3-(3-methyl-5-tert-butyl-4-hydroxyphenyl)propionate, Ethyl 3-(3-tert-butyl-4-hydroxy-5-methylphenyl)propionate, Ethyl 3-(3'-methyl-5'-tert-butyl-4'-hydroxyphenyl)propionate, Ethyl β-(5-tert-butyl-4-hydroxy-3-methylphenyl)propionate
IUPAC-namn ethyl 3-(3-tert-butyl-4-hydroxy-5-methylphenyl)propanoate
LEDER CCOC(=O)CCc1cc(C)c(O)c(c1)C(C)(C)C
Specifikationer
Kemiskt namn eller material Ethyl 3-(3-tert-Butyl-4-hydroxy-5-methylphenyl)propionate
Kvantitet 500 mg
InChI formel InChI=1S/C16H24O3/c1-6-19-14(17)8-7-12-9-11(2)15(18)13(10-12)16(3,4)5/h9-10,18H,6-8H2,1-5H3
Formel vikt 264.1725