Läs mer
Ethyl 3-(4-Pyridyl)propiolate, TRC
Produktkod. 30373251
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30373251 100 mg
1 options
Produktkod. 30373251 Leverantör Toronto Research Chemicals Leverantörsnummer TRCE939943100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Ethyl 3-(4-Pyridyl)propiolate
CAS 66869-71-4
Molekylformel C10 H9 N O2
Kvantitet 100 mg
Synonym Ethyl 3-(4-Pyridinyl)-2-propynoate, Ethyl 4-Pyridinylpropiolate, Pyridin-4-ylpropynoic Acid Ethyl Ester
InChI formel InChI=1S/C10H9NO2/c1-2-13-10(12)4-3-9-5-7-11-8-6-9/h5-8H,2H2,1H3
LEDER CCOC(=O)C#Cc1ccncc1
IUPAC-namn ethyl 3-(4-pyridyl)prop-2-ynoate
Molekylvikt (g/mol) 175.184
Formel vikt 175.0633