Läs mer
Ethyl 3-Aminobutanoate, TRC
Produktkod. 30337664
Change view
Kvantitet:
500 mg
Produktkod. Kvantitet
30337664 500 mg
1 options
Produktkod. 30337664 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB433860500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Ethyl 3-Aminobutanoate
CAS 5303-65-1
Molekylformel C6H13NO2
Kvantitet 500 mg
Synonym 3-Aminobutanoic Acid Ethyl Ester, 3-Aminobutyric Acid Ethyl Ester, 3-Methyl-beta-alanine Ethyl Ester, Ethyl 3-aminobutyrate, Ethyl beta-aminobutyrate, NSC 77084, [2-(Ethoxycarbonyl)-1-methylethyl]amine, beta-Aminobutyric Acid Ethyl Ester
InChI formel InChI=1S/C6H13NO2/c1-3-9-6(8)4-5(2)7/h5H,3-4,7H2,1-2H3
LEDER CCOC(=O)CC(C)N
IUPAC-namn ethyl 3-aminobutanoate
Molekylvikt (g/mol) 131.17
Formel vikt 131.0946