missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3-furoate, 99%, Thermo Scientific Chemicals

Catalog Number p-130143
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
614-98-2
C7H8O3
140.14
MFCD00005348
LOFDXZJSDVCYAS-UHFFFAOYSA-N
ethyl 3-furoate, ethyl 3-furancarboxylate, unii-i8q0yn919k, 3-furancarboxylic acid, ethyl ester, 3-furancarboxylic acid ethyl ester, ethyl 3furoate, ethyl3-furancarboxylate, ethyl3-furoate, pubchem19999, acmc-1bczk
69201
ethyl furan-3-carboxylate
CCOC(=O)C1=COC=C1
This item is not returnable. View return policy
614-98-2
C7H8O3
140.14
MFCD00005348
LOFDXZJSDVCYAS-UHFFFAOYSA-N
ethyl 3-furoate, ethyl 3-furancarboxylate, unii-i8q0yn919k, 3-furancarboxylic acid, ethyl ester, 3-furancarboxylic acid ethyl ester, ethyl 3furoate, ethyl3-furancarboxylate, ethyl3-furoate, pubchem19999, acmc-1bczk
69201
ethyl furan-3-carboxylate
CCOC(=O)C1=COC=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Density 1.1100g/mL
Flash Point 59°C
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4600 to 1.462
Quantity 1 g
Beilstein 18,4052
Specific Gravity 1.11
Formula Weight 140.14
Percent Purity 99%
Chemical Name or Material Ethyl 3-furoate, 99%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'