Läs mer
Ethyl 3-Hydroxyphenylacetate, TRC
Produktkod. 30379763
Change view
Kvantitet:
2 g
Produktkod. Kvantitet
30379763 2 g
1 options
Produktkod. 30379763 Leverantör Toronto Research Chemicals Leverantörsnummer TRCE9190752G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Ethyl 3-Hydroxyphenylacetate
CAS 22446-38-4
Molekylformel C10 H12 O3
Kvantitet 2 g
Synonym Benzeneacetic acid, 3-hydroxy-, ethyl ester, Acetic acid, (m-hydroxyphenyl)-, ethyl ester (8CI), Ethyl 3-hydroxybenzeneacetate, (3-Hydroxyphenyl)acetic acid ethyl ester, 2-(3-Hydroxyphenyl)acetic acid ethyl ester, Ethyl 2-(3-hydroxyphenyl)acetate, Ethyl 3-hydroxyphenylacetate
InChI formel InChI=1S/C10H12O3/c1-2-13-10(12)7-8-4-3-5-9(11)6-8/h3-6,11H,2,7H2,1H3
LEDER CCOC(=O)Cc1cccc(O)c1
IUPAC-namn ethyl 2-(3-hydroxyphenyl)acetate
Molekylvikt (g/mol) 180.2
Formel vikt 180.0786