Learn More
Ethyl 3-Mercaptopropanoate, TRC
Product Code. 30386694
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
Product Code. Quantity Unit Size
30386694 25 g 25g
1 options
Product Code. 30386694 Supplier Toronto Research Chemicals Supplier No. TRCE93234025G
 Restricted Item
Beräknad leverans: 09-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Ethyl 3-Mercaptopropanoate
CAS 5466-06-8
Molecular Formula C5H10O2S
Quantity 25 g
Synonym 3-Mercapto-propanoic Acid Ethyl Ester, 3-Mercapto-propionic Acid Ethyl Ester, 3-Mercaptopropanoic Acid Ethyl Ester, 3-Mercaptopropionic Acid Ethyl Ester, EPM, EPM (thiol), Ethyl 3-Mercaptopropanoate, Ethyl 3-Mercaptopropionate, Ethyl beta-Mercaptopropionate, NSC 25754, beta-Mercaptopropionic Acid Ethyl Ester
InChI Formula InChI=1S/C5H10O2S/c1-2-7-5(6)3-4-8/h8H,2-4H2,1H3
SMILES CCOC(=O)CCS
IUPAC Name ethyl 3-sulfanylpropanoate
Molecular Weight (g/mol) 134.2
Formula Weight 134.0402