Learn More
Ethyl 3-(Methyl-d3-mercapto)propionate, TRC
Product Code. 30385793
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
Product Code. Quantity Unit Size
30385793 5 mg 5mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30385793 Supplier Toronto Research Chemicals Supplier No. TRCE9251025MG
 Restricted Item
Beräknad leverans: 10-12-2026
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Ethyl 3-(Methyl-d3-mercapto)propionate
CAS 1020719-41-8
Molecular Formula C6 2H3 H9 O2 S
Quantity 5 mg
InChI Formula InChI=1S/C6H12O2S/c1-3-8-6(7)4-5-9-2/h3-5H2,1-2H3/i2D3
SMILES [2H]C([2H])([2H])SCCC(=O)OCC
IUPAC Name ethyl 3-(trideuteriomethylsulfanyl)propanoate
Molecular Weight (g/mol) 151.24
Formula Weight 151.0746