missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl 3-Oxo-4-aza-5alpha-androst-1-ene-17beta-carboxylate, Mikromol™
Description
Ethyl 3-Oxo-4-aza-5alpha-androst-1-ene-17beta-carboxylate
Specifications
Specifications
| Kemiskt namn eller material | Ethyl 3-Oxo-4-aza-5alpha-androst-1-ene-17beta-carboxylate |
| CAS | 157307-36-3 |
| Analyt- eller komponentnamn | Ethyl 3-Oxo-4-aza-5alpha-androst-1-ene-17beta-carboxylate |
| Frakt skick | Room Temperature |
| Molekylformel | C21 H31 N O3 |
| Certifikat och efterlevnad | ISO 9001 |
| Synonym | Dutasteride EP Impurity C, ethyl (4aR,4bS,6aS,7S,9aS,9bS,11aR)-4a,6a-dimethyl-2-oxo-2,4a,4b,5,6,6a,7,8,9,9a,9b,10,11,11a-tetradecahydro-1H-indeno[5,4-f]quinoline-7-carboxylate, 1H-Indeno[5,4-f]quinoline-7-carboxylic acid, 2,4a,4b,5,6,6a,7,8,9,9a,9b,10,11,11a-tetradecahydro-4a,6a-dimethyl-2-oxo-, ethyl ester, (4aR,4bS,6aS,7S,9aS,9bS,11aR)-, 4-Azaandrost-1-ene-17-carboxylic acid, 3-oxo-, ethyl ester, (5α,17β)-, 1H-Indeno[5,4-f]quinoline, 4-azaandrost-1-ene-17-carboxylic acid deriv., Ethyl 3-oxo-4-aza-5α-androst-1-ene-17β-carboxylate, 3-Oxo-4-aza-5α-androst-1-ene-17β-carboxylic Acid Ethyl Ester, Dutasteride Imp C (EP), Dutasteride Ethyl Ester, Dutasteride Impurity C |
| InChI formel | InChI=1S/C21H31NO3/c1-4-25-19(24)16-7-6-14-13-5-8-17-21(3,12-10-18(23)22-17)15(13)9-11-20(14,16)2/h10,12-17H,4-9,11H2,1-3H3,(H,22,23)/t13-,14-,15-,16+,17+,20-,21+/m0/s1 |
| LEDER | CCOC(=O)[C@H]1CC[C@H]2[C@@H]3CC[C@H]4NC(=O)C=C[C@]4(C)[C@H]3CC[C@]12C |
| IUPAC-namn | ethyl (1S,3aS,3bS,5aR,9aR,9bS,11aS)-9a,11a-dimethyl-7-oxo-1,2,3,3a,3b,4,5,5a,6,9b,10,11-dodecahydroindeno[5,4-f]quinoline-1-carboxylate |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?