missing translation for 'onlineSavingsMsg'
Läs mer

Ethyl 3-(p-Menthane-3-carboxamido)acetate, TRC

Produktkod. 30133603
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
1 g
5 g
10 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30133603 10 g
30136217 5 g
30011940 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 30133603 Leverantör Toronto Research Chemicals Leverantörsnummer TRCE68489010G

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 68489-14-5
Molekylformel C15 H27 N O3
Molekylvikt (g/mol) 269.38
Synonym N-[[(1R,2S,5R)-5-Methyl-2-(1-methylethyl)cyclohexyl]carbonyl]glycine Ethyl Ester, CPS 337, FEMA 4309;
IUPAC-namn ethyl 2-[[(1R,2S,5R)-5-methyl-2-propan-2-ylcyclohexanecarbonyl]amino]acetate
LEDER CCOC(=O)CNC(=O)[C@@H]1C[C@H](C)CC[C@H]1C(C)C

Specifications

Kemiskt namn eller material Ethyl 3-(p-Menthane-3-carboxamido)acetate
Anteckningar om renhetsgrad HPLC
InChI formel InChI=1S/C15H27NO3/c1-5-19-14(17)9-16-15(18)13-8-11(4)6-7-12(13)10(2)3/h10-13H,5-9H2,1-4H3,(H,16,18)/t11-,12+,13-/m1/s1
Kvantitet 10 g
Formel vikt 269.199
Procent renhet >95%
Frakt skick Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.