missing translation for 'onlineSavingsMsg'
Läs mer

Ethyl 4-(2,3,4-Trimethoxybenzyl)piperazine-1-carboxylate, Mikromol™

Produktkod. 30052438
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30052438 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 30052438 Leverantör Mikromol Leverantörsnummer MM0539.09

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Ethyl 4-(2,3,4-Trimethoxybenzyl)piperazine-1-carboxylate
TRUSTED_SUSTAINABILITY

Specifikationer

Kemiskt namn eller material Ethyl 4-(2,3,4-Trimethoxybenzyl)piperazine-1-carboxylate
CAS 53531-01-4
Analyt- eller komponentnamn Ethyl 4-(2,3,4-Trimethoxybenzyl)piperazine-1-carboxylate
Frakt skick Room Temperature
Molekylformel C17 H26 N2 O5
Certifikat och efterlevnad ISO 9001
Synonym Trimetazidine Dihydrochloride Imp. H (EP), Trimetazidine Imp. H (EP), Ethyl 4-(2,3,4-Trimethoxybenzyl)piperazine-1-carboxylate, Trimetazidine Dihydrochloride Impurity H, Trimetazidine Impurity H
InChI formel InChI=1S/C17H26N2O5/c1-5-24-17(20)19-10-8-18(9-11-19)12-13-6-7-14(21-2)16(23-4)15(13)22-3/h6-7H,5,8-12H2,1-4H3
LEDER CCOC(=O)N1CCN(Cc2ccc(OC)c(OC)c2OC)CC1
IUPAC-namn ethyl 4-[(2,3,4-trimethoxyphenyl)methyl]piperazine-1-carboxylate
Molekylvikt (g/mol) 338.4
Kvantitet 100 mg
Formel vikt 338.1842 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.