Learn More
Ethyl 4-[2-(5-Chloro-2-methoxybenzamido)ethyl]benzene Sulfonamide Carbamate, TRC
Product Code. 30135634
Change view
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30135634 1 g 1g
30011575 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30135634 Supplier Toronto Research Chemicals Supplier No. TRCE9019601G
 Restricted Item
Beräknad leverans: 09-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 14511-59-2
Molecular Formula C19 H21 Cl N2 O6 S
Molecular Weight (g/mol) 440.9
IUPAC Name ethyl N-[4-[2-[(5-chloro-2-methoxybenzoyl)amino]ethyl]phenyl]sulfonylcarbamate
SMILES CCOC(=O)NS(=O)(=O)c1ccc(CCNC(=O)c2cc(Cl)ccc2OC)cc1
Specifications
Quantity 1 g
InChI Formula InChI=1S/C19H21ClN2O6S/c1-3-28-19(24)22-29(25,26)15-7-4-13(5-8-15)10-11-21-18(23)16-12-14(20)6-9-17(16)27-2/h4-9,12H,3,10-11H2,1-2H3,(H,21,23)(H,22,24)
Formula Weight 440.0809
Chemical Name or Material Ethyl 4-[2-(5-Chloro-2-methoxybenzamido)ethyl]benzene sulfonamide carbamate