Läs mer
Ethyl 4-(4-Fluorophenyl)-6-isopropyl-2-[methyl(methylsulfonyl)amino]pyrimidine-5-carboxylate, Mikromol™
Produktkod. 30050392
Change view
Kvantitet:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30050392 100 mg
30050109 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30050392 Leverantör Mikromol Leverantörsnummer MM0936.09
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Ethyl 4-(4-Fluorophenyl)-6-isopropyl-2-[methyl(methylsulfonyl)amino]pyrimidine-5-carboxylate
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Ethyl 4-(4-Fluorophenyl)-6-isopropyl-2-[methyl(methylsulfonyl)amino]pyrimidine-5-carboxylate
CAS 147118-30-7
Analyt- eller komponentnamn Ethyl 4-(4-Fluorophenyl)-6-isopropyl-2-[methyl(methylsulfonyl)amino]pyrimidine-5-carboxylate
Frakt skick Room Temperature
Molekylformel C18 H22 F N3 O4 S
Certifikat och efterlevnad ISO 17025
Synonym 5-Pyrimidinecarboxylic acid, 4-(4-fluorophenyl)-6-(1-methylethyl)-2-[methyl(methylsulfonyl)amino]-, ethyl ester, Ethyl 4-(4-fluorophenyl)-6-isopropyl-2-[methyl(methylsulfonyl)amino]pyrimidine-5-carboxylate
InChI formel InChI=1S/C18H22FN3O4S/c1-6-26-17(23)14-15(11(2)3)20-18(22(4)27(5,24)25)21-16(14)12-7-9-13(19)10-8-12/h7-11H,6H2,1-5H3
LEDER CCOC(=O)c1c(nc(nc1c2ccc(F)cc2)N(C)S(=O)(=O)C)C(C)C
IUPAC-namn ethyl 4-(4-fluorophenyl)-2-[methyl(methylsulfonyl)amino]-6-propan-2-ylpyrimidine-5-carboxylate
Molekylvikt (g/mol) 395.45
Kvantitet 100 mg
Formel vikt 395.1315 g/mol