Läs mer
Ethyl 4,5-Bis(2-methoxyethoxy)-2-nitrobenzoate, TRC
Produktkod. 30376010
Change view
Kvantitet:
1 g
Produktkod. Kvantitet
30376010 1 g
1 options
Produktkod. 30376010 Leverantör Toronto Research Chemicals Leverantörsnummer TRCE9000801G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Ethyl 4,5-Bis(2-methoxyethoxy)-2-nitrobenzoate
CAS 179688-26-7
Molekylformel C15H21NO8
Kvantitet 1 g
Synonym 4,5-Bis(2-methoxyethoxy)-2-nitro-benzoic Acid Ethyl Ester
InChI formel InChI=1S/C15H21NO8/c1-4-22-15(17)11-9-13(23-7-5-20-2)14(24-8-6-21-3)10-12(11)16(18)19/h9-10H,4-8H2,1-3H3
LEDER CCOC(=O)c1cc(OCCOC)c(OCCOC)cc1[N+](=O)[O-]
Molekylvikt (g/mol) 343.33
Formel vikt 343.1267