Läs mer
Ethyl 4,6-Dichloropyridazine-3-carboxylate, TRC
Produktkod. 30375342
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30375342 100 mg
1 options
Produktkod. 30375342 Leverantör Toronto Research Chemicals Leverantörsnummer TRCE903353100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Ethyl 4,6-Dichloropyridazine-3-carboxylate
CAS 679406-03-2
Molekylformel C7 H6 Cl2 N2 O2
Kvantitet 100 mg
Synonym 4,6-Dichloropyridazine-3-carboxylic Acid Ethyl Ester, 4,6-Dichloro-3-pyridazinecarboxylic Acid Ethyl Ester
InChI formel InChI=1S/C7H6Cl2N2O2/c1-2-13-7(12)6-4(8)3-5(9)10-11-6/h3H,2H2,1H3
LEDER CCOC(=O)c1nnc(Cl)cc1Cl
IUPAC-namn ethyl 4,6-dichloropyridazine-3-carboxylate
Molekylvikt (g/mol) 221.041
Formel vikt 219.981