missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Ethyl 4,6-dihydroxynicotinate, 95%

Brand:  Alfa Aesar™ H63842.06

Additional Details : CAS Number : 6975-44-6 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15450759

  • 2853.88 SEK / 5g


Estimated Shipment date
from Supplier 12-03-2018
Add to basket

Specifications

Chemical Identifiers

Ethyl 4,6-dihydroxynicotinate
183.16
4,6-Dihydroxynicotinic acid ethyl ester; Ethyl 4,6-dihydroxypyridine-3-carboxylate
6975-44-6
MFCD00834976

Specifications

Ethyl 4,6-dihydroxynicotinate
6975-44-6
95%
MFCD00834976
5g
Solid
183.16
4,6-Dihydroxynicotinic acid ethyl ester; Ethyl 4,6-dihydroxypyridine-3-carboxylate
Videos
SDS