Learn More
Ethyl 4-acetyl-5-oxohexanoate, 98%
Product Code. 11368456
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11368456 25 g 25g
11358456 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11368456 Supplier Thermo Scientific Alfa Aesar Supplier No. L17496.14
 Restricted Item
I lager
Add to basket
CAS: 2832-10-2 | C11H21BrO2 | 265.19 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 2832-10-2
MDL Number MFCD00009211
Specifications
Density 1.067
Boiling Point 153°C to 154°C (19 mmHg)
Flash Point >110°C (230°F)
Molecular Formula C10H16O4
Refractive Index 1.4545
Quantity 25 g
Beilstein 1785189
Synonym ethyl 4,4-diacetylbutyrate, 4-acetyl-5-oxohexanoic acid ethyl ester, 4,4-diacetylbutyric acid ethyl ester, ethyl 4-acetyl-5-oxohexanoate, 4-acetyl-5-oxo-hexanoic acid ethyl ester, hexanoic acid,4-acetyl-5-oxo-, ethyl ester, hexanoic acid, 4-acetyl-5-oxo-, ethyl ester
InChI Key CVPCFJRJUCNUJG-VQHVLOKHSA-N
SMILES CCOC(=O)CCC(C(=O)C)C(=O)C
IUPAC Name ethyl 4-acetyl-5-oxohexanoate
Molecular Weight (g/mol) 200.23
PubChem CID 2733934
Formula Weight 200.24
Percent Purity 98%
Chemical Name or Material Ethyl 4-acetyl-5-oxohexanoate