Läs mer
Ethyl 4-Amino-2-hydroxybenzoate (Ethyl Aminosalicylate), Mikromol™
Produktkod. 30050385
Change view
Kvantitet:
25 mg
Produktkod. Kvantitet
30050385 25 mg
1 options
Produktkod. 30050385 Leverantör Mikromol Leverantörsnummer MM1645.020025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Ethyl 4-Amino-2-hydroxybenzoate (Ethyl Aminosalicylate)
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Ethyl 4-Amino-2-hydroxybenzoate (Ethyl Aminosalicylate)
CAS 6059-17-2
Analyt- eller komponentnamn Ethyl 4-Amino-2-hydroxybenzoate (Ethyl Aminosalicylate)
Frakt skick Room Temperature
Molekylformel C9 H11 N O3
Certifikat och efterlevnad ISO 9001
Synonym 4-Amino-2-hydroxybenzoic acid ethyl ester, Salicylic acid, 4-amino-, ethyl ester (6CI,7CI,8CI), Ethyl 2-hydroxy-4-aminobenzoate, Ethyl 4-amino-2-hydroxybenzoate, Ethyl 4-aminosalicylate, Ethyl p-aminosalicylate, p-Aminosalicylic acid ethyl ester
InChI formel InChI=1S/C9H11NO3/c1-2-13-9(12)7-4-3-6(10)5-8(7)11/h3-5,11H,2,10H2,1H3
LEDER CCOC(=O)c1ccc(N)cc1O
IUPAC-namn ethyl 4-amino-2-hydroxybenzoate
Molekylvikt (g/mol) 181.19
Kvantitet 25 mg
Formel vikt 181.0739 g/mol