missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Ethyl 4-Aminopentanoate Hydrochloride, TRC
Specifications
Specifications
| Kemiskt namn eller material | Ethyl 4-Aminopentanoate Hydrochloride |
| CAS | 119396-97-3 |
| Molekylformel | C7H15NO2 . HCl |
| Kvantitet | 50 mg |
| Synonym | Pentanoic acid, 4-amino-, ethyl ester, hydrochloride (1:1) (ACI), Pentanoic acid, 4-amino-, ethyl ester, hydrochloride (9CI), 4-Aminopentanoic acid ethyl ester hydrochloride, 4-Aminovaleric acid ethyl ester hydrochloride, Ethyl 4-aminovalerate hydrochloride, Ethyl 4-aminopentanoate hydrochloride |
| InChI formel | InChI=1S/C7H15NO2.ClH/c1-3-10-7(9)5-4-6(2)8;/h6H,3-5,8H2,1-2H3;1H |
| LEDER | Cl.CCOC(=O)CCC(C)N |
| IUPAC-namn | ethyl 4-aminopentanoate;hydrochloride |
| Molekylvikt (g/mol) | 181.66 |
| Formel vikt | 181.087 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?