Läs mer
Ethyl 4-Biphenylacetate, TRC
Produktkod. 30375494
Change view
Kvantitet:
50 mg
100 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30375494 50 mg
30387280 100 mg
30387801 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30375494 Leverantör Toronto Research Chemicals Leverantörsnummer TRCE90020850MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 14062-23-8
Molekylformel C16 H16 O2
Molekylvikt (g/mol) 240.3
Synonym [1,1'-Biphenyl]-4-acetic acid ethyl ester, 4-Biphenylacetic acid, ethyl ester (8CI), Acetic acid, (4-biphenylyl)-, ethyl ester (7CI), Diatec, Ethyl 4-biphenylylacetate, Ethyl p-biphenylylacetate, Felbinac ethyl ester, LM 001
IUPAC-namn ethyl 2-(4-phenylphenyl)acetate
LEDER CCOC(=O)Cc1ccc(cc1)c2ccccc2
Specifikationer
Kemiskt namn eller material Felbinac Ethyl Ester
Kvantitet 50 mg
InChI formel InChI=1S/C16H16O2/c1-2-18-16(17)12-13-8-10-15(11-9-13)14-6-4-3-5-7-14/h3-11H,2,12H2,1H3
Formel vikt 240.115