Läs mer
Ethyl 4-(Butylamino)benzoate(4-(Butylamino)benzoic Acid Ethyl Ester), TRC
Produktkod. 30343955
Change view
Kvantitet:
1 g
Produktkod. Kvantitet
30343955 1 g
1 options
Produktkod. 30343955 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB6907051G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Ethyl 4-(Butylamino)benzoate(4-(Butylamino)benzoic Acid Ethyl Ester)
CAS 94-32-6
Molekylformel C13 H19 N O2
Kvantitet 1 g
Synonym Benzoic acid, 4-(butylamino)-, ethyl ester, Benzoic acid, p-(butylamino)-, ethyl ester (6CI,7CI,8CI), Ethyl p-butylaminobenzoate, 4-(Butylamino)benzoic acid ethyl ester, Ethyl 4-(butylamino)benzoate
InChI formel InChI=1S/C13H19NO2/c1-3-5-10-14-12-8-6-11(7-9-12)13(15)16-4-2/h6-9,14H,3-5,10H2,1-2H3
LEDER CCCCNc1ccc(cc1)C(=O)OCC
IUPAC-namn ethyl 4-(butylamino)benzoate
Molekylvikt (g/mol) 221.3
Formel vikt 221.1416