missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 4-chloro-8-cyanoquinoline-3-carboxylate, 97%, Thermo Scientific™

Catalog Number p-7077329
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
77173-67-2
C13H9ClN2O2
260.68
MHOFAUCLSGVKOH-UHFFFAOYSA-N
ethyl4-chloro-8-cyanoquinoline-3-carboxylate, ethyl 4-chloro-8-cyano-3-quinolinecarboxylate, pubchem19877, 4-chloro-8-cyanoquinoline-3-carboxylicacidethyleste, 4-chloro-8-cyanoquinoline-3-carboxylic acid ethyl ester, ethyl 4-chloro-8-cyanoquinoxaline-3-carboxylate
3761179
ethyl 4-chloro-8-cyanoquinoline-3-carboxylate
CCOC(=O)C1=C(C2=CC=CC(=C2N=C1)C#N)Cl
This item is not returnable. View return policy
77173-67-2
C13H9ClN2O2
260.68
MHOFAUCLSGVKOH-UHFFFAOYSA-N
ethyl4-chloro-8-cyanoquinoline-3-carboxylate, ethyl 4-chloro-8-cyano-3-quinolinecarboxylate, pubchem19877, 4-chloro-8-cyanoquinoline-3-carboxylicacidethyleste, 4-chloro-8-cyanoquinoline-3-carboxylic acid ethyl ester, ethyl 4-chloro-8-cyanoquinoxaline-3-carboxylate
3761179
ethyl 4-chloro-8-cyanoquinoline-3-carboxylate
CCOC(=O)C1=C(C2=CC=CC(=C2N=C1)C#N)Cl
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 260.68
Percent Purity 97%
Chemical Name or Material Ethyl 4-chloro-8-cyanoquinoline-3-carboxylate
compliance-icons
Product Identifier
  • Ethyl 4-chloro-8-cyanoquinoline-3-carboxylate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'