Learn More
Ethyl 4-Hydroxy-2-methyl-2H-1,2-benzothiazine-3-carboxylate 1,1-Dioxide, Mikromol™
Product Code. 17930406
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17930406 25 mg 25mg
17992126 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17930406 Supplier Mikromol Supplier No. MM0084.030025
 Restricted Item
Beräknad leverans: 30-11-2026
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Ethyl 4-Hydroxy-2-methyl-2H-1,2-benzothiazine-3-carboxylate 1,1-Dioxide
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material ethyl 4-hydroxy-2-methyl-2H-1,2-benzothiazine-3-carboxylate 1,1-dioxide
CAS 24683-26-9
Analyte or Component Names ethyl 4-hydroxy-2-methyl-2H-1,2-benzothiazine-3-carboxylate 1,1-dioxide
Shipping Condition Room Temperature
Molecular Formula C12 H13 N O5 S
Certifications/Compliance ISO 17025
Synonym Meloxicam Imp. A (EP), Piroxicam Imp. K (EP), Meloxicam USP Related Compound A, Meloxicam USP RC A, Ethyl 4-Hydroxy-2-methyl-2H-1,2-benzothiazine-3-carboxylate 1,1-Dioxide, Meloxicam Related Compound A, Meloxicam Impurity A, Piroxicam Impurity K
InChI Formula InChI=1S/C12H13NO5S/c1-3-18-12(15)10-11(14)8-6-4-5-7-9(8)19(16,17)13(10)2/h4-7,14H,3H2,1-2H3
SMILES CCOC(=O)C1=C(O)c2ccccc2S(=O)(=O)N1C
IUPAC Name ethyl 4-hydroxy-2-methyl-1,1-dioxo-1λ^{6},2-benzothiazine-3-carboxylate
Molecular Weight (g/mol) 283.3
Quantity 25 mg
Formula Weight 283.0514 g/mol