Läs mer
Ethyl 4-Oxo-4-(3-pyridyl)butanoate, TRC
Produktkod. 30374792
Change view
Kvantitet:
50 mg
Produktkod. Kvantitet
30374792 50 mg
1 options
Produktkod. 30374792 Leverantör Toronto Research Chemicals Leverantörsnummer TRCE92540050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Ethyl 4-Oxo-4-(3-pyridyl)butanoate
CAS 59086-27-0
Molekylformel C11 H13 N O3
Kvantitet 50 mg
Synonym 3-Pyridinebutanoic acid, γ-oxo-, ethyl ester, Ethyl γ-oxo-3-pyridinebutanoate, Ethyl 3-nicotinoyl propionate, Ethyl 4-oxo-4-(3-pyridyl)butanoate, Ethyl 4-oxo-4-(pyridin-3-yl)butanoate, 4-Oxo-4-(3-pyridyl)butanoic acid ethyl ester, γ-Oxo-3-pyridinebutanoic acid ethyl ester
InChI formel InChI=1S/C11H13NO3/c1-2-15-11(14)6-5-10(13)9-4-3-7-12-8-9/h3-4,7-8H,2,5-6H2,1H3
LEDER CCOC(=O)CCC(=O)c1cccnc1
IUPAC-namn ethyl 4-oxo-4-pyridin-3-ylbutanoate
Molekylvikt (g/mol) 207.23
Formel vikt 207.0895