Läs mer
Ethyl 4-Phenylacetoacetate, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | Ethyl 4-Phenylacetoacetate |
| CAS | 718-08-1 |
| Molekylformel | C12 H14 O3 |
| Kvantitet | 1 g |
| Synonym | beta-oxo-Benzenebutanoic acid ethyl ester, 4-Phenyl-acetoacetic acid ethyl ester, 3-Oxo-4-phenylbutanoic acid ethyl ester, 4-Phenyl-3-oxobutanoic acid ethyl ester, 4-Phenylacetoacetic acid ethyl ester, Ethyl 3-keto-4-phenylbutyrate, Ethyl 3-Oxo-4-phenylbutanoate, Ethyl 3-oxo-4-phenylbutyrate, Ethyl 3-Oxobenzenebutanoate, Ethyl 4-phenyl-3-oxobutanoate, Ethyl beta-Oxobenzenebutanoate, NSC 15691 |
| InChI formel | InChI=1S/C12H14O3/c1-2-15-12(14)9-11(13)8-10-6-4-3-5-7-10/h3-7H,2,8-9H2,1H3 |
| LEDER | O=C(OCC)CC(=O)CC=1C=CC=CC1 |
| IUPAC-namn | ethyl 3-oxo-4-phenyl-butanoate |
| Molekylvikt (g/mol) | 206.23776 |
| Formel vikt | 206.0943 |