Läs mer
Ethyl 5-(1-Methyl-5-nitro-benzimidazol-2-yl)pentanoate, Mikromol™
Produktkod. 30051173
Change view
Kvantitet:
25 mg
100 mg
4 x 25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30051173 100 mg
30049119 25 mg
30460293 4 x 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30051173 Leverantör Mikromol Leverantörsnummer MM0154.53
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Ethyl 5-(1-Methyl-5-nitro-benzimidazol-2-yl)pentanoate
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Ethyl 5-(1-Methyl-5-nitro-benzimidazol-2-yl)pentanoate
CAS 3543-76-8
Analyt- eller komponentnamn Ethyl 5-(1-Methyl-5-nitro-benzimidazol-2-yl)pentanoate
Frakt skick Room Temperature
Molekylformel C15 H19 N3 O4
Certifikat och efterlevnad ISO 9001
Synonym 1H-Benzimidazole-2-pentanoic acid, 1-methyl-5-nitro-, ethyl ester, 2-Benzimidazolevaleric acid, 1-methyl-5-nitro-, ethyl ester (7CI,8CI)
InChI formel InChI=1S/C15H19N3O4/c1-3-22-15(19)7-5-4-6-14-16-12-10-11(18(20)21)8-9-13(12)17(14)2/h8-10H,3-7H2,1-2H3
LEDER CCOC(=O)CCCCc1nc2cc(ccc2n1C)[N+](=O)[O-]
IUPAC-namn ethyl 5-(1-methyl-5-nitrobenzimidazol-2-yl)pentanoate
Molekylvikt (g/mol) 305.33
Kvantitet 100 mg
Formel vikt 305.1376 g/mol