missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Ethyl 5-bromo-1-indoleacetate, 97%

Brand:  Alfa Aesar™ H63224.06

Additional Details : CAS Number : 726174-45-4 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15499798

  • 6791.60 SEK / 5g


Estimated Shipment date
from Supplier 18-02-2019
Add to basket

Specifications

Chemical Identifiers

726174-45-4
282.137
ASRCALVKWKRKTR-UHFFFAOYSA-N
43152067
CCOC(=O)CN1C=CC2=C1C=CC(=C2)Br
C12H12BrNO2
MFCD11205770
5-bromo-1h-indol-1-yl-acetic acid ethyl ester, 1h-indole-1-acetic acid, 5-bromo-, ethyl ester, ethyl 2-5-bromoindol-1-yl acetate, ethyl 5-bromo-1h-indol-1-yl acetate, 5-bromo-indol-1-yl-acetic acid ethyl ester
ethyl 2-(5-bromoindol-1-yl)acetate

Specifications

Ethyl 5-bromo-1-indoleacetate
726174-45-4
MFCD11205770
5-bromo-1h-indol-1-yl-acetic acid ethyl ester, 1h-indole-1-acetic acid, 5-bromo-, ethyl ester, ethyl 2-5-bromoindol-1-yl acetate, ethyl 5-bromo-1h-indol-1-yl acetate, 5-bromo-indol-1-yl-acetic acid ethyl ester
CCOC(=O)CN1C=CC2=C1C=CC(=C2)Br
282.137
282.13
97%
5g
C12H12BrNO2
9195073
ASRCALVKWKRKTR-UHFFFAOYSA-N
ethyl 2-(5-bromoindol-1-yl)acetate
43152067
Solid
Videos
SDS