missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl 5-bromo-2-methylnicotinate, 97%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
Ethyl 5-bromo-2-methylnicotinate is used as a pharmaceutical intermediate.
Solubility
Slightly soluble in water.
Notes
Store in cool, dry place in tightly closed container. Store away from oxidizing agent.
Ethyl 5-bromo-2-methylnicotinate is used as a pharmaceutical intermediate.
Solubility
Slightly soluble in water.
Notes
Store in cool, dry place in tightly closed container. Store away from oxidizing agent.
Specifications
Color | Yellow |
Quantity | 250 mg |
Synonym | ethyl 5-bromo-2-methylnicotinate, 5-bromo-2-methyl-nicotinic acid ethyl ester, 5-bromo-2-methyl-3-pyridinecarboxylic acid ethyl ester, 5-bromo-2-methylnicotinic acid ethyl ester, 3-pyridinecarboxylic acid, 5-bromo-2-methyl-, ethyl ester, ethyl5-bromo-2-methylnicotinate, ethyl 5-bromo-2-methyl-pyridine-3-carboxylate |
Solubility Information | Slightly soluble in water. |
Formula Weight | 244.09 |
Percent Purity | 97% |
Physical Form | Solid |
Chemical Name or Material | Ethyl 5-bromo-2-methylnicotinate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'