Learn More
Ethyl 5-bromothiophene-2-carboxylate, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
25 g
Description
Ethyl 5-bromothiophene-2-carboxylate is used as an intermediate in organic syntheses
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Ethyl 5-bromothiophene-2-carboxylate is used as an intermediate in organic syntheses
Solubility
Slightly soluble in water
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Ethyl 5-bromothiophene-2-carboxylate is used as an intermediate in organic syntheses
Solubility
Slightly soluble in water
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications
Boiling Point | 94°C to 96°C (4 mmHg) |
Quantity | 1 g |
Solubility Information | Slightly soluble in water |
Formula Weight | 235.1 |
Percent Purity | 99% |
Chemical Name or Material | Ethyl 5-bromothiophene-2-carboxylate |