missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 6-chloronicotinate, 97%, Thermo Scientific™

Catalog Number 15244378
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
49608-01-7
C8H8ClNO2
185.61
MFCD00082739
ILDJJTQWIZLGPO-UHFFFAOYSA-N
2799611
ethyl 6-chloropyridine-3-carboxylate
CCOC(=O)C1=CC=C(Cl)N=C1
This item is not returnable. View return policy
49608-01-7
C8H8ClNO2
185.61
MFCD00082739
ILDJJTQWIZLGPO-UHFFFAOYSA-N
2799611
ethyl 6-chloropyridine-3-carboxylate
CCOC(=O)C1=CC=C(Cl)N=C1
Melting Point 26.0°C to 30.0°C
Boiling Point 98.0°C (1.5 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Quantity 5 g
Synonym ethyl 6-chloronicotinate, ethyl 6-chloro-3-pyridinecarboxylate, ethyl6-chloronicotinate, 6-chloro-nicotinic acid ethyl ester, 6-chloronicotinic acid ethyl ester, ethyl 6-chloro nicotinate, ethyl 2-chloro-5-pyridinecarboxylate, 3-pyridinecarboxylic acid, 6-chloro-, ethyl ester, pubchem1415, pubchem15179
Formula Weight 185.61
Percent Purity 97%
Chemical Name or Material Ethyl 6-chloronicotinate
compliance-icons
Product Identifier
  • Ethyl 6-chloronicotinate, 97%
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'