Läs mer
Ethyl 8-Bromooctanoate, TRC
Produktkod. 30379217
Change view
Kvantitet:
500 mg
Produktkod. Kvantitet
30379217 500 mg
1 options
Produktkod. 30379217 Leverantör Toronto Research Chemicals Leverantörsnummer TRCE900845500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Ethyl 8-Bromooctanoate
CAS 29823-21-0
Molekylformel C10H19BrO2
Kvantitet 500 mg
Synonym 8-Bromocaprylic Acid Ethyl Ester, 8-Bromooctanoic Acid Ethyl Ester, NSC 100182
InChI formel InChI=1S/C10H19BrO2/c1-2-13-10(12)8-6-4-3-5-7-9-11/h2-9H2,1H3
LEDER CCOC(CCCCCCCBr)=O
IUPAC-namn ethyl 8-bromooctanoate
Molekylvikt (g/mol) 251.16
Formel vikt 250.06