Läs mer
Ethyl a-Bromoethylglyoxalate, TRC
Produktkod. 30374356
Change view
Kvantitet:
5 g
Produktkod. Kvantitet
30374356 5 g
1 options
Produktkod. 30374356 Leverantör Toronto Research Chemicals Leverantörsnummer TRCE9003005G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Ethyl alpha-Bromoethylglyoxalate
CAS 57332-84-0
Molekylformel C6 H9 Br O3
Kvantitet 5 g
Synonym 3-Bromo-2-oxobutanoic Acid Ethyl Ester, 3-Bromo-2-oxobutyric Acid Ethyl Ester, Ethyl 3-bromo-2-oxobutanoate, Ethyl 3-bromo-2-oxobutyrate, 3-Bromo-2-oxo-butanoic Acid Ethyl Ester
InChI formel InChI=1S/C6H9BrO3/c1-3-10-6(9)5(8)4(2)7/h4H,3H2,1-2H3
LEDER CCOC(=O)C(=O)C(C)Br
IUPAC-namn ethyl 3-bromo-2-oxobutanoate
Molekylvikt (g/mol) 209.04
Formel vikt 207.9735