Läs mer
Ethyl Acetate-13C2, TRC
Produktkod. 30376343
Change view
Kvantitet:
25 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30376343 25 mg
30373385 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30376343 Leverantör Toronto Research Chemicals Leverantörsnummer TRCE67835325MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 84508-45-2
Molekylformel C213C2H8O2
Molekylvikt (g/mol) 90.09
Synonym Acetic Acid Ehyl Ester-13C2, Acetic Ether-13C2, Acetidin-13C2, Acetoxyethane-13C2, EtOAc-13C2, Ethyl Ethanoate-13C2, NSC 70930-13C2, Vinegar Naphtha-13C2
LEDER [13CH3][13C](OCC)=O
Specifikationer
Kemiskt namn eller material Ethyl Acetate-13C2
Kvantitet 25 mg
InChI formel InChI=1S/C4H8O2/c1-3-6-4(2)5/h3H2,1-2H3/i2+1,4+1