Läs mer
Ethyl Acetylsalicylate, Mikromol™
Produktkod. 30050181
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30050181 100 mg
1 options
Produktkod. 30050181 Leverantör Mikromol Leverantörsnummer MM0133.13
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Ethyl Acetylsalicylate
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Ethyl Acetylsalicylate
CAS 529-68-0
Analyt- eller komponentnamn Ethyl Acetylsalicylate
Frakt skick Room Temperature
Molekylformel C11 H12 O4
Certifikat och efterlevnad ISO 17025
Synonym 2-(Acetyloxy)-benzoic acid ethyl ester, Acetylsalicylic acid, ethyl ester (5CI), Salicylic acid, ethyl ester, acetate (8CI), 2-(Acetyloxy)benzoic acid ethyl ester, 2-Carbethoxyphenyl acetate, Ethyl 2-acetoxybenzoate, Ethyl acetylsalicylate, Ethyl o-acetoxybenzoate
InChI formel InChI=1S/C11H12O4/c1-3-14-11(13)9-6-4-5-7-10(9)15-8(2)12/h4-7H,3H2,1-2H3
LEDER CCOC(=O)c1ccccc1OC(=O)C
IUPAC-namn ethyl 2-acetyloxybenzoate
Molekylvikt (g/mol) 208.21
Kvantitet 100 mg
Formel vikt 208.0736 g/mol